missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RC Rat anti-Mouse, Oligo, Clone: DNL-1.9, BD™ AbSeq ™
Rat Monoclonal Antibody
Marke: BD Biosciences 940491
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CD45RC Monoclonal antibody specifically detects CD45RC in Mouse samples. It is validated for Single Cell 3' SequencingSpezifikation
CD45RC | |
Monoclonal | |
Oligo | |
Ptprc; Cd45; CD45R; L-CA; Ly-5; Lyt-4; T200 | |
70Z/3 mouse pre-B cell line | |
RUO | |
Mouse | |
IgG2a κ |
Single Cell 3' Sequencing | |
Clone DNL-1.9 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |