Learn More
CD49b (Integrin alpha 2) Monoclonal Antibody (HMa2), FITC, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
175.00 €
Spezifikation
Antigen | CD49b (Integrin alpha 2) |
---|---|
Klon | HMa2 |
Konzentration | 0.5 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15268109
|
Affymetrix eBioscience
11-0491-82 |
100 μg |
175.00 €
100 Mikrogramm |
Verfügbar ab: 10-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Description: The HMalpha2 monoclonal antibody reacts with mouse CD49b, the 150 kDa integrin alpha2 subunit. The complex of CD49b non-covalently associated with integrin beta1 (CD29), also known as VLA-2, is a receptor for collagen and laminin. This complex is expressed by some CD4^+ T cells, IEL, NK cells, platelets and epithelial cells. Applications Reported: The HM alpha2 antibody has been reported for use in flow cytometric analysis. Applications Tested: The HM alpha2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD49b (Integrin alpha 2, Integrin alpha 2 chain, VLA-2 alpha chain, HM alpha 2) is a member of the integrin family. It is a glycoprotein with molecular weight of 150 kD, and it complexes with CD29 (Integrin beta 1) to form the heterodimeric integrin VLA-2 (integrin alpha 2 beta 1, or GPIa/IIa) complex. VLA-2 is an extracellular receptor for laminin, collagen, and fibronectin, and interaction with its ligands results in the activation of intracellular signaling pathways. It has reported roles in VEGF-induced angiogenesis in vivo, as well as adhesion and lymphocyte activation. CD49b is expressed by NK cells, NK-T cells, monocytes, platelets, and epithelial cells. It is also expressed on adaptive immune cells such as T and B cells, specifically on a subset of CD4+ T cells in the spleen, on intraepithelial and lamina propria lymphocytes in the intestine, as well as on a population of peripheral CD4+ type 1 T regulatory (Tr1) cells that co-express LAG-3.Spezifikation
CD49b (Integrin alpha 2) | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
alpha 2 subunit of VLA-2 receptor; BDPLT9; BR; CD49 antigen-like family member B; CD49B; Collagen receptor; DX5; GPIa; HPA-5; human platelet alloantigen system 5; integrin alpha 2; integrin alpha-2; integrin subunit alpha 2; integrin, alpha 2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); Itga2; platelet antigen Br; platelet glycoprotein GPIa; platelet membrane glycoprotein Ia; very late activation protein 2 receptor, alpha-2 subunit; VLA-2; VLA2 receptor; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha; VLAA2 | |
Itga2 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Itga2 |
HMa2 | |
Flow Cytometry | |
FITC | |
Armenian Hamster | |
Mouse | |
Q62469 | |
16398 | |
IgG | |
Affinity chromatography | |
Antibody |
For Research Use Only.