Learn More
CD5 Monoclonal Antibody (L17F12), PE-eFluor™ 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
148.00 € - 349.00 €
Spezifikation
Antigen | CD5 |
---|---|
Klon | L17F12 |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16331074
|
Affymetrix eBioscience
61-0058-42 |
100 Tests |
349.00 €
100 Stück |
Verfügbar ab: 07-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16321074
|
Affymetrix eBioscience
61-0058-41 |
25 Tests |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Description: The L17F12 monoclonal antibody reacts with human CD5, a 67 kDa protein expressed by a majority of thymocytes and mature T cells and a subset of B cells. Signaling through the CD5 molecule activates T cells and binding of CD5 to its ligand on B cells, CD72, and plays an important role in T-B interaction and proliferation. Applications Reported: This L17F12 antibody has been reported for use in flow cytometric analysis. Applications Tested: This L17F12 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PE-eFluor 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red). Please make sure that your instrument is capable of detecting this fluorochrome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222-49) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No.
CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter's Syndrome.Spezifikation
CD5 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P06127 | |
IgG2a κ | |
4° C, store in dark, DO NOT FREEZE! |
L17F12 | |
Flow Cytometry | |
PE-eFluor 610 | |
Mouse | |
Human | |
921 | |
Primary | |
Antibody |