Learn More
CD56 (NCAM) Mouse, anti-Human, PE, Clone: HCD56, STEMCELL Technologies
Mouse Monoclonal Antibody
Marke: Stemcell Technologies 60021PE
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- The HCD56 antibody reacts with CD56 (NCAM), a member of the immunoglobulin (Ig) super family containing 5 Ig-like domains and two fibronectin type-3 domains in its extracellular portion
- Multiple isoforms have been identified, including transmembrane and GPI-anchored forms
- CD56 is subject to several types of post-translational modification, including addition of a polysialic acid moiety to the 5th Ig-like domain
- The ∼140 kDa isoform of CD56 is considered the prototypic cell surface marker on human NK cells
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ HLA Chimerism Buffy Coat CD56 Positive Selection Kit (Catalog #17875) and EasySep™ Human CD56 Positive Selection Kit II (Catalog #17855); partial blocking may be observed, as well as EasySep™ Human CD2 Positive Selection Kit II (Catalog #17883) and EasySep™ Human NK Cell Enrichment Kit (Catalog #19055)
Spezifikation
| CD56 (NCAM) | |
| Monoclonal | |
| PE | |
| 100 Tests | |
| Human | |
| NK Cells |
| CyTOF, Flow Cytometry, Immunohistochemistry | |
| HCD56 | |
| Mouse | |
| Primary | |
| IgG1 κ |
Berichtigung von Produktinhalten
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.