missing translation for 'onlineSavingsMsg'
Learn More

CD59 Mouse anti-Human, FITC, Clone: P282 (H19), BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 560954

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15870629

  • 221.00 € / 1 Stück
Verfügbar ab: 10-06-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The p282 (HI9) monoclonal antibody specifically binds to CD59, a 19 kDa glycosylphosphatidylinositol (GPI)-anchored glycoprotein, expressed on hematopoietic and non-hematopoietic cells. Because of its role in interaction with complement activated products, CD59 has been termed membrane-attack-complex-inhibitory factor (MACIF), homologus restriction factor (HRF20), membrane inhibitor of reactive lysis (MIRL) and Protectin. It inhibits the cytolytic activity of complement by binding to C8 and C9, thereby blocking the assembly of the membrane attack complex. CD59 also participates in spontaneous T-cell/erythrocyte adhesion, interacts with CD2 and plays a role in T-cell activation.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Spezifikation

Spezifikation

CD59
Monoclonal
FITC
Mouse
Affinity Purified
RUO
Human
IgG2a κ
Flow Cytometry
p282 (H19)
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human Erythrocytes
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.