missing translation for 'onlineSavingsMsg'
Learn More

CD64 Monoclonal Antibody (X54-5/7.1), APC, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Affymetrix eBioscience 17-0641-80

80.00 EUR gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15903204

  • 120.00 € / 25 Mikrogramm
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Description: This X54-5/7.1 monoclonal antibody recognizes allelic variants a and b of mouse CD64, a high affinity Fc binding receptor also known as Fc-gammaR1 or Fcgr1. Alloform a is present in the following mouse strains: C57BL/6, BALB/c, DBA/2, and alloform b in strains: C3H/HeJ, CBA/J, NZW, SJL/J, 129/SvJ. In addition this antibody has been reported positive in strains: AKR, ALR, BUB, C58, CE, HRS,MRL, MON, NZB, NZO, NZW, PL, SJL, ST and SWR. This clone X54-5/7.1 will not recognize the allelic variant d, present in NOD/Lt mice. This antibody has been reported to stain RAW264.7 cell line. Applications Reported: This X54-5/7.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This X54-5/7.1 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser

CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.
Spezifikation

Spezifikation

CD64
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P26151
Fcgr1
BALB/c mouse CD64-human IgG Fc fusion protein
25 μg
Primary
Mouse
Antibody
IgG1 κ
Flow Cytometry
X54-5/7.1
APC
Fcgr1
AI323638; AV092959; CD64; CD64 antigen; CD64A; Fc fragment of IgG high affinity Ib receptor; Fc fragment of IgG receptor Ia; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc fragment of IgG, high affinity Ib, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc receptor, IgG, high affinity I; Fcg1; Fc-gamma receptor 1; Fc-gamma receptor I A1; Fc-gamma RI; Fc-gamma RIA; FcgammaRI; FcgammaRIa; Fcgr1; FCGR1A; Fcgr1b; FcgRI; FCRI; FLJ18345; high affinity IgG Fc receptor, subunit beta; high affinity immunoglobulin gamma Fc receptor I; high-affinity immunoglobulin gamma Fc receptor I; IGFR1; IgG Fc receptor I; IgG high affinity Fc receptor; IGGHAFC; RP11-196G18.2
Mouse
Affinity chromatography
RUO
14129
4° C, store in dark, DO NOT FREEZE!
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote