Learn More
CD7 Antibody (CD7/8496R), Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP3-20469-20ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Spezifikation
| CD7 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
| Recombinant fragment (around aa 72-175) of human CD7 protein (exact sequence is proprietary) | |
| 20 μg | |
| Cytokine Research, Signal Transduction | |
| 924 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| CD7/8496R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |