missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD74 Monoclonal Antibody (5-329), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marke: Affymetrix eBioscience 62-0748-41
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Mouse Monoclonal Antibody
CD74 Monoclonal antibody specifically detects CD74 in Human samples. It is validated for Flow CytometrySpezifikation
CD74 | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P04233 | |
CD74 | |
Affinity chromatography | |
RUO | |
972 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
5-329 | |
Super Bright 436 | |
CD74 | |
Cd74; CD74 antigen; CLIP; DHLAG; H-2 class II histocompatibility antigen gamma chain; HLA class II histocompatibility antigen gamma chain; HLADG; HLA-DR antigens-associated invariant chain; Ia antigen-associated invariant chain; Ia-GAMMA; II; INVG34; MHC class II-associated invariant chain; p33 | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |