Learn More
CD8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Marke:  Novus Biologicals NBP2-29475
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 18749133
  • 575.00 € / 0.10 Milligramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
CD8 Polyclonal specifically detects CD8 in Human, Mouse, Porcine samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
CD8
CD8, CD8a molecule, LEU2, p32
32 kDa
0.1 mg
Flow Cytometry, Immunofluorescence, Immunoprecipitation
Recombinant fragment from human CD8 C-terminal cytoplasmic domain of alpha chain (exact sequence is proprietary). (Uniprot: P01732)
Protein A purified
Pig