Learn More
CD8a Rat anti-Mouse, PerCP, Clone: 53-6.7, BD
Rat Monoclonal Antibody
171.00 € - 410.00 €
Spezifikation
Antigen | CD8a |
---|---|
Klon | 53-6.7 |
Konzentration | 0.2mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15890759
|
BD Biosciences
561092 |
25 μg |
171.00 €
1 Stück |
Verfügbar ab: 23-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15856948
|
BD Biosciences
553036 |
0.1 mg |
410.00 €
1 Stück |
Verfügbar ab: 23-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
The 53-6.7 antibody monoclonal antibody specifically binds to the 38 kDa α and 34 kDa α chains of the CD8 differentiation antigen (Ly-2 or Lyt-2) of all mouse strains tested. The CD8 α and α chains (CD8a) form heterodimers with the CD8 β chain (CD8b, Ly-3, or Lyt-3) on the surface of most thymocytes. A subpopulation of mature T lymphocytes (i.e., MHC class I-restricted T cells, including most T suppressor/cytotoxic cells) expresses almost exclusively the CD8 αβ heterodimer (the αα chains arise from alternatively spliced messengers of a single CD8a gene. The longer α form associates with p56 [lck] via a CXCP motif in its cytoplasmic domain, which it shares with CD4, but not with CD8b. The truncated α chain is unable to associate with p56 [lck], and it may function to attenuate the CD8-mediated costimulatory signal during intrathymic T-cell maturation. In vivo and in vitro treatment with 53-6.7 mAb has reportedly been effective at depleting CD8+ peripheral T lymphocytes. The 53-6.7 antibody has also been reported to cross-reaact with CD8 α- and α-like polypeptides on subsets of thymic and peripheral lymphocytes in the Egyptian toad, Bufo regularis.
Spezifikation
CD8a | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
IgG2a κ | |
Affinity Purified | |
Cd8a; CD8 alpha chain; Ly-2; Lyt2; Lyt-2; Ly-35; Ly-B |
53-6.7 | |
Flow Cytometry | |
PerCP | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.