missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD90 Monoclonal Antibody (OX-7)

Mouse Monoclonal Antibody
276.00 € - 277.00 €
Spezifikation
| Antigen | CD90 |
|---|---|
| Klon | OX-7 |
| Konzentration | 1 mg/mL |
| Inhalt und Lagerung | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Anwendungen | Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunocytochemistry, Western Blot |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
11535512
|
Invitrogen™
MA180651 |
250 μg |
277.00 €
250 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
11590003
![]() |
Invitrogen™
MA181491 |
100 μg |
276.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Rat CD90 antibody, clone OX-7 recognizes rat and CD90, also known as Thy1. Mouse anti Rat CD90 antibody, clone OX-7 recognizes rat and CD90, also known as Thy1.
CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. CD90 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90 can be used as a marker for a variety of stem cells and for the axonal processes of mature neurons. Structural study of CD90 led to the foundation of the Immunoglobulin superfamily, of which it is the smallest member, and led to the first biochemical description and characterization of a vertebrate GPI anchor. Diseases associated with CD90 dysfunction include nasopharyngeal carcinoma and thymoma.Spezifikation
| CD90 | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P01830, P01831 | |
| 100339279, 100379563, 21838, 24832 | |
| Rat Thy1 antigen. | |
| Antibody |
| OX-7 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Mouse, Rat, Rabbit, Guinea Pig | |
| Thy1 | |
| CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
| Thy1 | |
| Primary | |
| Protein A |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts