missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90 Mouse anti-Human, Porcine, Rat, Clone: 2D7D11, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Spezifikation
| Antigen | CD90 |
|---|---|
| Klon | 2D7D11 |
| Konzentration | 1 mg/mL |
| Anwendungen | Western Blot, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
16848945
|
Proteintech
66766-1-IG-20UL |
20 μL |
145.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
16838945
|
Proteintech
66766-1-IG-150UL |
150 μL |
410.00 €
150 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
THY1 encodes a cell surface glycoprotein and member of the immunoglobulin superfamily of proteins. The encoded protein is involved in cell adhesion and cell communication in numerous cell types, but particularly in cells of the immune and nervous systems. The encoded protein is widely used as a marker for hematopoietic stem cells. This gene may function as a tumor suppressor in nasopharyngeal carcinoma. Alternative splicing results in multiple transcript variants.Spezifikation
| CD90 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CD90, CDw90, FLJ33325, Thy 1 antigen, Thy 1 cell surface antigen, Thy 1 membrane glycoprotein, THY1 | |
| THY1 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 2D7D11 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Rat, Porcine | |
| P01830, P04216 | |
| 100271931, 24832, 7070 | |
| CD90 Fusion Protein Ag25603 | |
| Primary | |
| -20°C | |
| THY1 |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts