missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD94 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
N.V.
Spezifikation
Antigen | CD94 |
---|---|
Klon | 131412 |
Konzentration | LYOPH |
Verdünnung | Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready |
Anwendungen | Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF |
Beschreibung
CD94 Monoclonal specifically detects CD94 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Spezifikation
CD94 | |
LYOPH | |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
CD94, CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KLRD1, KP43, NK cell receptor | |
KLRD1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
131412 | |
Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
3824 | |
BaF3 mouse pro-B cell line transfected with human CD94 and NKG2A | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Recognizes human CD94 both in its homodimeric form and as a heterodimer with either NKG2A or NKG2C. |
For Research Use Only