Learn More
CD95 Mouse anti-Human, PE-Cy5, Clone: DX2, BD
Mouse Monoclonal Antibody
240.00 € - 415.00 €
Spezifikation
Antigen | CD95 |
---|---|
Klon | DX2 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | PE-Cyanine5 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15821529
|
BD Biosciences
561977 |
25 Tests |
240.00 €
1 Stück |
Verfügbar ab: 07-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15879768
|
BD Biosciences
559773 |
100 Tests |
415.00 €
1 Stück |
Verfügbar ab: 07-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
The DX2 monoclonal antibody specifically binds to the human Fas antigen (also called APO-1). This 45 kDa transmembrane cell surface molecule was designated as CD95 at the Fifth HLDA Workshop. Fas is a member of the TNF-receptor superfamily and is also known as Tumor necrosis factor receptor superfamily member 6 (TNFRSF6). It is expressed on a variety of normal and neoplastic cells including activated T and B lymphocytes and some undifferentiated thymocytes. The Fas/CD95 antigen is a polypeptide that plays a role in the programmed sequence of events leading to cell death, termed apoptosis. The DX2 clone specifically reacts with murine L cells, murine L1210 leukemia cells and murine P815 mastocytoma cells transfected with human Fas cDNA but not with untransfected parental cell lines. Crosslinking CD95 with DX2 antibody delivers an apoptotic signal indicating that DX2 recognizes a functional epitope of the CD95 antigen.
Spezifikation
CD95 | |
Flow Cytometry | |
PE-Cyanine5 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
APO-1; FAS; TNFRSF6; APT1; ALPS1A; FAS1; FASTM; FASLG receptor |
DX2 | |
Monoclonal | |
Mouse | |
Baboon, Canine, Cynomolgus, Human, Porcine, Rhesus Monkey | |
Human CD95-transfected L Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.