Learn More
Cdc20B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562867
Additional Details : Netto-Gewicht : 0.02000kg
Description
Immunogen sequence: EASGSVLKTP PEKETLTLGS RKEQLKTPSK GISETSNSAL HFCKAPHAMD RDWKESVASK GQKCLKQLFV TQNVVQQANG KMQLCEQS Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 29%.
Cdc20B Polyclonal antibody specifically detects Cdc20B in Human samples. It is validated for Immunohistochemistry.Specifications
Cdc20B | |
Polyclonal | |
Unconjugated | |
CDC20B | |
CDC20 cell division cycle 20 homolog B; CDC20B; CDC20-like protein; cell division cycle 20 homolog B; cell division cycle 20B; cell division cycle protein 20 homolog B; G6VTS76519 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
166979 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86Y33 | |
CDC20B | |
Recombinant protein corresponding to Human Cdc20B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only