missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC25C Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16485-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
M-phase inducer phosphatase 3is anenzymethat in humans is encoded by theCDC25Cgene. This gene is highly conserved during evolution and it plays a key role in the regulation of cell division. The encoded protein is a tyrosine phosphatase and belongs to the Cdc25 phosphatase family. It directs dephosphorylation of cyclin B-bound CDC2 (CDK1) and triggers entry into mitosis. Also, it is thought to suppress p53-induced growth arrest. Multiple alternatively spliced transcript variants of this gene have been described, however, the full-length nature of many of them is not known.Spezifikation
CDC25C | |
Polyclonal | |
Unconjugated | |
CDC25C | |
CDC25, CDC25C, M phase inducer phosphatase 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
995 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P30307 | |
CDC25C | |
CDC25C Fusion Protein Ag9607 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |