Learn More
CDH9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552394
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GIITVKQNLD FENQMLYTLR VDASNTHPDP RFLHLGPFKD TAVVKISVED IDEPPVFTKV SYLIEVDEDV KEGSIIGQVT AYDPDARNNL IKYSVDRHTD MDRI Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
CDH9 (cadherin-9) is a type II classical cadherin from the cadherin superfamily, integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. The extracellular domain consists of 5 subdomains, each containing a cadherin motif, and appears to determine the specificity of the protein's homophilic cell adhesion activity. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins.Spezifikation
CDH9 | |
Polyclonal | |
Unconjugated | |
CDH9 | |
Cadherin; cadherin 9; cadherin 9, type 2 (T1-cadherin); cadherin 9, type 2, T1-cadherin; cadherin-9; CDH9; T1-cadherin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1007 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULB4 | |
CDH9 | |
Recombinant protein corresponding to Human CDH9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only