missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17182-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene belongs to the CDK subfamily of the Ser/Thr protein kinase family. The CDK subfamily members are highly similar to the gene products of S. cerevisiae cdc28, and S. pombe cdc2, and are known to be essential for cell cycle progression. This kinase has been shown to play a role in cellular proliferation and its function is limited to cell cycle G2-M phase. Several alternatively spliced transcripts encoding different isoforms, some of which use non-AUG (CUG) translation initiation codon, have been found for this gene.Spezifikation
CDK10 | |
Polyclonal | |
Unconjugated | |
CDK10 | |
CDK10, cyclin dependent kinase 10, PISSLRE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
234854, 361434, 8558 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15131, Q3UMM4, Q4KM47 | |
Cdk10 | |
CDK10 Fusion Protein Ag10935 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |