missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK6 Monoclonal Antibody (DCS83.1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA112301
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
CDK6 functions as a cell-cycle initiator protein. This protein is coexpressed and copurified with CyclinD1. They are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression. CDK6 is essential for cell proliferation within the dentate gyrus of the hippocampus and the cuventricular zone of the lateral ventricles. Mutations in the gene results in microcephaly 12, primary, autosomal recessive (MCPH12).Spezifikation
CDK6 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.08% sodium azide | |
Q00534 | |
CDK6 | |
Recombinant Cdk6 protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
DCS83.1 | |
Unconjugated | |
CDK6 | |
5830411I20; AI504062; CDK6; CDKN6; Cell division protein kinase 6; CR2 protein kinase; Crk2; cyclin dependent kinase 6; cyclin-dependent kinase 6; MCPH12; MGC59692; PLSTIRE; Serine/threonine-protein kinase PLSTIRE | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
1021 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |