missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKAL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546800
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: QEDSKPQDRH FVRKDVVPKV RRRNTQKYLQ EEENSPPSDS TIPGIQKIWI
CDKAL1 Polyclonal antibody specifically detects CDKAL1 in Human samples. It is validated for Western Blot.Spezifikation
CDKAL1 | |
Polyclonal | |
Unconjugated | |
Cdkal1 | |
1190005B03Rik; 6620401C13Rik; CDK5 regulatory subunit associated protein 1 like 1; CDK5 regulatory subunit associated protein 1-like 1; CDK5 regulatory subunit-associated protein 1-like 1; CDKAL; CDKAL1; Threonylcarbamoyladenosine tRNA methylthiotransferase; tRNA-t(6)A37 methylthiotransferase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54901 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5VV42 | |
Cdkal1 | |
Synthetic peptide directed towards the N-terminal of human CDKAL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |