Learn More
CDKN2AIPNL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560564
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RLDQLLSLSM VWANHLFLGC SYNKDLLDKV MEMADGIEVE DLPQFTTRSE LMK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
CDKN2AIPNL Polyclonal antibody specifically detects CDKN2AIPNL in Human samples. It is validated for Immunohistochemistry.Spezifikation
CDKN2AIPNL | |
Polyclonal | |
Unconjugated | |
CDKN2AIPNL | |
CDKN2A interacting protein N-terminal like; CDKN2A-interacting protein N-terminal-like protein; CDKN2AIP N-terminal-like protein; CDKN2AIPNL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91368 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96HQ2 | |
CDKN2AIPNL | |
Recombinant protein corresponding to Human CDKN2AIPNL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only