Learn More
CDV3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556253
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KAVTKDEDEW KELEQKEVDY SGLRVQAMQI SSEKEEDDNE KRQDPGDNWE E Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.
The function of this protein remains unknown.Spezifikation
CDV3 | |
Polyclonal | |
Unconjugated | |
CDV3 | |
2510010F10Rik; C230084J24Rik; C79446; carnitine deficiency-associated gene expressed in ventricle 3; carnitine deficiency-associated gene expressed in ventricle 3 homolog; carnitine deficiency-associated protein 3; Cdv3; CDV3 homolog; H41; Protein CDV3; protein CDV3 homolog; TPP36; Tyrosine-phosphorylated protein 36 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55573 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKY7 | |
CDV3 | |
Recombinant protein corresponding to Human CDV3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only