missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP41 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA539090
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
This gene encodes a centrosomal and microtubule-binding protein which is predicted to have two coiled-coil domains and a rhodanese domain. In human retinal pigment epithelial cells the protein localized to centrioles and cilia. Mutations in this gene have been associated with Joubert Syndrome 15; an autosomal recessive ciliopathy and neurological disorder. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Mar 2012].Spezifikation
CEP41 | |
Polyclonal | |
Unconjugated | |
CEP41 | |
JBTS15, TSGA14, centrosomal protein 41 kDa, centrosomal protein of 41 kDa, testis specific protein A14, testis specific, 14, testis-specific gene A14 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
95681 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9BYV8 | |
CEP41 | |
A synthetic peptide derived from the internal region of human CEP41 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |