missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerebellin 4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA539096
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Cerebellin (CER), which was originally isolated from rat cerebellum, is a hexadecapeptide derived from a larger precursor called Cerebellin 1, also designated precerebellin 1 or Cbln1. Four propeptides, Cerebellin 1, Cerebellin 2 (Cbln2), Cerebellin 3 (Cbln3) and Cerebellin 4 (Cbln4), comprise the precerebellin subfamily within the C1q protein family. Cerebellin family members act as transneuronal regulators of synapse development and synaptic plasticity in various brain regions. Cerebellin and its metabolite, des-Ser(1)Cer, are also expressed in several extra-cerebellar tissues, including adrenal gland. Cerebellin 1, 2 and 3 assemble into homomeric and heteromeric complexes, thereby influencing each other′s degradation and secretion. Cerebellin 3 is not able to form homomeric complexes, and can only be secreted upon forming a heteromeric complex with Cerebellin 1. Decreased concentrations of Cerebellin have been found in the brain of patients with olivopontocerebellar atrophy (OPCA) and Shy-Drager syndrome, suggesting a role for Cerebellin in the pathology of these diseases.Spezifikation
Cerebellin 4 | |
Polyclonal | |
Unconjugated | |
CBLN4 | |
Cerebellin-4, Cerebellin-like glycoprotein 1, CBLN4, CBLNL1, UNQ718/PRO1382, Cerebellin4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140689, 228942 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NTU7, Q8BME9 | |
CBLN4 | |
A synthetic peptide derived from the internal region of human CBLN4 | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG |