missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CES1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16912-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the carboxylesterase large family. The family members are responsible for the hydrolysis or transesterification of various xenobiotics, such as cocaine and heroin, and endogenous substrates with ester, thioester, or amide bonds. They may participate in fatty acyl and cholesterol ester metabolism, and may play a role in the blood-brain barrier system. This enzyme is the major liver enzyme and functions in liver drug clearance. Mutations of this gene cause carboxylesterase 1 deficiency. Three transcript variants encoding three different isoforms have been found for this gene.Spezifikation
CES1 | |
Polyclonal | |
Unconjugated | |
CES1 | |
ACAT, Brain carboxylesterase hBr1, CE1, CEH, CES1, CES2, Cocaine carboxylesterase, Egasyn, HMSE, HMSE1, Liver carboxylesterase 1, PCE 1, REH, Retinyl ester hydrolase, Serine esterase 1, SES1, TGH, Triacylglycerol hydrolase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1066 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P23141 | |
CES1 | |
CES1 Fusion Protein Ag10567 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |