missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CFHL2 (Human) Recombinant Protein
Human CFHL2 full-length ORF ( AAH22283, 19 a.a. - 270 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00003080-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: EAMFCDFPKINHGILYDEEKYKPFSQVPTGEVFYYSCEYNFVSPSKSFWTRITCAEEGWSPTPKCLRLCFFPFVENGHSESSGQTHLEGDTVQIICNTGYRLQNNENNISCVERGWSTPPKCRSTISAEKCGPPPPIDNGDITSFLLSVYAPGSSVEYQCQNLYQLEGNNQITCRNGQWSEPPKCLDPCVISQEIMEKYNIKLKWTNQQKLYSRTGDIVEFVCKSGYHPTKSHSFRAMCQNGKLVYPSCEEK
Spezifikation
AAH22283 | |
complement factor H-related 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CFHR2 | |
GST |
3080 | |
Wheat germ expression system | |
25 μg | |
CFHL2, FHR2, HFL3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |