missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFHR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA568399
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Might be involved in complement regulation. Can associate with lipoproteins and may play a role in lipid metabolism.Spezifikation
CFHR2 | |
Polyclonal | |
Unconjugated | |
CFHR2 | |
CFHL2; CFHR2; complement factor H related 2; complement factor H-related 2; complement factor H-related protein 2; DDESK59; factor H-related gene 2; FHR2; FHR-2; H factor (complement)-like 3; h factor-like 3; H factor-like protein 2; HFL3 | |
Rabbit | |
affinity chromatography | |
RUO | |
3080 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P36980 | |
CFHR2 | |
Synthetic protein corresponding to residues near the C terminal of human CFHR2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |