missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CGA Recombinant Rabbit Monoclonal Antibody (0I8N3)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA558122
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: APDVQDCPEC TLQENPFFSQ PGAPILQCMG CCFSRAYPTP LRSKKTMLVQ KNVTSESTCC VAKSYNRVTV MGGFKVENHT ACHCSTCYYH KS.
The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family.
Spezifikation
| CGA | |
| Recombinant Monoclonal | |
| 1.04 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P01215 | |
| CGA | |
| Recombinant protein (or fragment) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 0I8N3 | |
| Unconjugated | |
| CGA | |
| aGSU; alphaGSU; alpha-GSU; alphaSU; Anterior pituitary glycoprotein hormones common subunit alpha; Cga; CG-ALPHA; Choriogonadotropin alpha chain; chorionic gonadotrophin subunit alpha; chorionic gonadotropin human; chorionic gonadotropin, alpha polypeptide; follicle-stimulating hormone alpha chain; follicle-stimulating hormone alpha subunit; follitropin alpha chain; FSHA; FSH-alpha; glycoprotein hormones alpha chain; glycoprotein hormones, alpha polypeptide; glycoprotein hormones, alpha subunit; GPHa; GPHA1; GPHalpha; HCG; human chorionic gonadotropin; LHA; LSH-alpha; luteinizing hormone alpha chain; Lutropin alpha chain; thyroid-stimulating hormone alpha chain; Thyrotropin alpha chain; Tsha; TSH-alpha | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1081 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur