missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGRP Monoclonal Antibody (18B7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA120287
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-20287 detects catacalcin in human samples.
This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
CGRP | |
Monoclonal | |
6.3 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P01258, P06881 | |
Calca | |
Fragments of human PCT conjugated with carrier protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
18B7 | |
Unconjugated | |
Calca | |
alpha-type CGRP; CA; Cal1; CAL6; CALC; Calc1; Calca; calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRP-I; Ct; Ctn; Katacalcin; KC; MGC126648; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; RAT calcitonin; RATCAL6 | |
Mouse | |
Protein A | |
RUO | |
796 | |
4° C | |
Liquid |