missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA537148
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 230-250 kDa. Purity is >95% by SDS-PAGE.
CHD5 Polyclonal antibody specifically detects CHD5 in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
CHD5 | |
Polyclonal | |
Unconjugated | |
CHD5 | |
4930532L22Rik; ATP-dependent helicase CHD5; AW060752; B230399N07Rik; CHD5; CHD-5; chromodomain helicase DNA binding protein 5; chromodomain-helicase-DNA-binding protein 5; KIAA0444 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26038, 269610, 691589 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
A2A8L1, D3ZD32, Q8TDI0 | |
CHD5 | |
A synthetic peptide corresponding to residues in human CHD5 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |