missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHEK1 (CHK1) Recombinant Human Protein
CHEK1 (CHK1) is a serine/threonine protein kinase.
Marke: Invitrogen™ PR3959B
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
CHEK1 (CHK1) is involved in preventing replication when DNA integrity is compromised.
Spezifikation
NP_001265 | |
50mM Tris, pH 7.5, 250mM NaCl, 0.05mM EDTA, 0.05% NP-40, 1mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
CHEK1 (CHK1) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
58.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CAMKL Kinase Family | |
Recombinant | |
Baculovirus |