Learn More
DiscoverX ChemiScreen™ CCK1 Cholecystokinin Receptor Stable Cell Line
Artikelnummer. 17315339
Encompass
Ansicht ändern
Click to view available options
Menge:
2 Fläschchen
Unit Size:
2 Ampullen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
17315339 2 Fläschchen 2 Ampullen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17315339 Supplier DiscoverX Supplier No. HTS184C
 Eingeschränkter Artikel
Verfügbar ab: 17-09-2026
Add to basket
Add to basket
The ChemiScreen™ CCK1 Cholecystokinin Receptor Stable Cell Line measures CCKAR (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.
The ChemiScreen™ CCK1 Cholecystokinin Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. cholecystokinin) based activation of CCKAR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CCKAR alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of CCKAR , the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Assay-Eingabe Calcium Flux
Zelllinie Chem-1
Zur Verwendung mit (Anwendung) Cell Lines
Primärer Agonist (Tyr[SO3]27) Cholecystokinin (26-33)
Menge 2 Fläschchen
Zieleintrag CCKAR
Zugriffsnummer AY322549