Learn More
DiscoverX ChemiScreen™ CCK1 Cholecystokinin Receptor Stable Cell Line
Description
The ChemiScreen™ CCK1 Cholecystokinin Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. cholecystokinin) based activation of CCKAR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CCKAR alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of CCKAR , the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay-Eingabe | Calcium Flux |
| Zelllinie | Chem-1 |
| Zur Verwendung mit (Anwendung) | Cell Lines |
| Primärer Agonist | (Tyr[SO3]27) Cholecystokinin (26-33) |
| Menge | 2 Fläschchen |
| Zieleintrag | CCKAR |
| Zugriffsnummer | AY322549 |