Learn More
DiscoverX ChemiScreen™ CCR7 Chemokine Receptor Stable Cell Line
Artikelnummer. 17384969
Encompass
Change view
Click to view available options
Menge:
2 Fläschchen
Artikelnummer. Menge
17384969 2 Fläschchen
1 options
Artikelnummer. 17384969
Marke: DiscoverX HTS012C
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
The ChemiScreen™ CCR7 Chemokine Receptor Stable Cell Line measures CCR7 (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.
The ChemiScreen™ CCR7 Chemokine Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CCL19) based activation of CCR7 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CCR7 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of CCR7 , the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells.
Spezifikation
Assay-Eingabe Calcium Flux
Zelllinie Chem-1
Zur Verwendung mit (Anwendung) Cell Lines
Primärer Agonist Recombinant Human MIP-3β (CCL19)
Menge 2 Fläschchen
Zieleintrag CCR7
Zugriffsnummer AY587876