Learn More
DiscoverX ChemiScreen™ GLP-1 Receptor Membrane Preparation
Click to view available options
Menge:
1 Fläschchen (200 Einheiten)
Beschreibung
The ChemiScreen™ GLP-1 Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of GLP1R surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach or to screen for GPCR activity using GTPγS assays. Increased binding affinities or GPCR activation levels will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.
Spezifikation
Spezifikation
| Assay-Eingabe | GLP-1 |
| Zur Verwendung mit (Anwendung) | Membrane Preps |
| Menge | 1 Fläschchen (200 Einheiten) |
| Zieleintrag | GLP1R |