Learn More
DiscoverX CHO-K1 CYSLTR2 Gq Cell Line
Artikelnummer. 17384739
Encompass
Ansicht ändern
Click to view available options
Menge:
2 Fläschchen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
17384739 2 Fläschchen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17384739 Lieferant DiscoverX Lieferanten-Nr. 951013C2
 Eingeschränkter Artikel
Verfügbar ab: 15-09-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
The CHO-K1 CYSLTR2 Gq Cell Line measures CYSLTR2 (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.
The CYSLTR2 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. leukotriene) based activation of CYSLTR2 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CYSLTR2 , a naturally Gq coupled GPCR. Upon activation of CYSLTR2 , the resulting calcium activity can be fluorescently assayed using the Calcium NoWash PLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells.
Spezifikation
Assay-Eingabe Calcium Signaling
Zelllinie CHO-K1
Zur Verwendung mit (Anwendung) Cell Lines
Primärer Agonist Leukotriene C4
Menge 2 Fläschchen
Zieleintrag CYSLTR2
Zugriffsnummer NM_020377.2