missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRM4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA567650
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Predicted to react with Mouse samples.
Muscarinic acetylcholine receptor M4 is an Acetylcholine Receptor. Unlike muscarinic receptor M2, the M4 receptor is not involved in smooth muscle contraction. The clinical relevance of this receptor is unknown. However, mouse studies have linked it to adenylyl cyclase inhibition. Muscarinic acetylcholine receptor M4 has been reported to be expressed in many regions of the rat brain, particularly the striatum and hippocampus. ESTs have been isolated from human breast and eye libraries.Spezifikation
CHRM4 | |
Polyclonal | |
Unconjugated | |
CHRM4 | |
acetylcholine receptor; acetylcholine receptor, muscarinic 4; AChR M4; cholinergic receptor; cholinergic receptor muscarinic 4; cholinergic receptor, muscarin 4; cholinergic receptor, muscarinic 4; CHRM4; Chrm-4; HM4; M4; M4R; Mm4 mAChR; muscarinic acetylcholine receptor 4; muscarinic acetylcholine receptor M4 | |
Rabbit | |
affinity chromatography | |
RUO | |
1132, 12672 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P08173, P32211 | |
CHRM4 | |
Synthesized peptide derived from human CHRM4. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |