missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRM5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533435
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%) Gibbon (94%) Chimpanzee, Marmoset (88%) Dog, Bovine, Panda, Horse (81%).
The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The clinical implications of this receptor are unknown; however, stimulation of this receptor is known to increase cyclic AMP levels.Spezifikation
CHRM5 | |
Polyclonal | |
Unconjugated | |
CHRM5 | |
acetylcholine receptor; acetylcholine receptor, muscarinic 5; AChR-M5; cholinergic receptor; cholinergic receptor muscarinic 5; cholinergic receptor, muscarinic 5; Chrm5; Chrm-5; HM5; m5; M5R; muscarinic acetylcholine receptor 5; muscarinic acetylcholine receptor M5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1133 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P08912 | |
CHRM5 | |
Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human CHRM5. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |