Learn More
CHRNB3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546827
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: YDGTMVDLIL INENVDRKDF FDNGEWEILN AKGMKGNRRD GVYSYPFITY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Mouse: 100%; Rabbit: 100%; Rat: 100%
CHRNB3 Polyclonal antibody specifically detects CHRNB3 in Human samples. It is validated for Western Blot.Spezifikation
CHRNB3 | |
Polyclonal | |
Unconjugated | |
Chrnb3 | |
5730417K16Rik; acetylcholine receptor; acetylcholine receptor, neuronal nicotinic, beta-3 subunit; acetylcholine receptor, nicotinic, beta 3 (neuronal); Acrb3; cholinergic receptor; cholinergic receptor nicotinic beta 3 subunit; cholinergic receptor, nicotinic beta 3; cholinergic receptor, nicotinic, beta 3 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 3; Chrnb3; I79_003985; Neuronal acetylcholine receptor subunit beta-3; neuronal nicotinic | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1142 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q05901 | |
Chrnb3 | |
Synthetic peptide directed towards the middle region of human CHRNB3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |