missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHST1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA536935
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 47 kDa. Purity is >95% by SDS-PAGE.
CHST1 Polyclonal antibody specifically detects CHST1 in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
CHST1 | |
Polyclonal | |
Unconjugated | |
CHST1 | |
2610008E20Rik; AW125896; C6ST; carbohydrate (chondroitin 6/keratan) sulfotransferase 1; carbohydrate (keratan sulfate Gal-6) sulfotransferase 1; carbohydrate sulfotransferase 1; chst1; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 1; Gst1; GST-1; hypothetical protein LOC445124; keratan sulfate Gal-6 sulfotransferase; Keratan sulfotransferase; KS6ST; KSGal6ST; KSST; N-acetylglucosamine-6-O-sulfotransferase 1; sulfo1; sulfotransferase1; zgc:100904; zgc:100904 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
295934, 76969, 8534 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O43916, Q5RJQ0, Q9EQC0 | |
CHST1 | |
Synthetic peptide corresponding to amino acids 126-171 of human CHST1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |