Learn More
CHST10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553002
Weitere Details : net-weight : 0.02000kg
Beschreibung
Immunogen sequence: PEVRKLPEEK HIPEELKPTG KELPDSQLVQ PLVYMERLEL IRNVCRDDAL KNLSHTPVSK FVLDRIFVCD KHKILFCQTP KVGNTQWKKV LIVLNGAFSS IEEIPENVVH DHEKNGLPRL SSFSDAEIQK RL Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.
This protein encoded by this gene transfers sulfate to the C-3 hydroxyl of terminal glucuronic acid of protein- and lipid-linked oligosaccharides. This protein was first identified as a sulfotransferase that acts on the human natural killer-1 (HNK-1) glycan; HNK-1 is a carbohydrate involved in neurodevelopment and synaptic plasticity.[provided by RefSeq, Feb 2011].Spezifikation
CHST10 | |
Polyclonal | |
Unconjugated | |
CHST10 | |
AI507003; AU041319; carbohydrate sulfotransferase 10; CHST10; HNK-1 sulfotransferase; HNK1ST; HNK-1ST; huHNK-1ST; raHNK-1ST; ST; sul-T | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9486 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43529 | |
CHST10 | |
Recombinant protein corresponding to Human CHST10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only