missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHST5 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547740
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) CHST1, rhCHST2, rmCHST5, and rmCHST7 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CHST5 Polyclonal antibody specifically detects CHST5 in Human samples. It is validated for Immunoprecipitation, Western Blot.Spezifikation
CHST5 | |
Polyclonal | |
Unconjugated | |
CHST5 | |
AI173964; carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 5; carbohydrate sulfotransferase 5; C-GlcNAc6ST; CHST5; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4; galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-alpha; GlcNAc6ST-3; Gn6st-3; GST4; GST-4; GST4-alpha; GST4-beta; hIGn6ST; I-GlcNAc6ST; I-GlcNAc-6-ST; intestinal GlcNAc-6-sulfotransferase; intestinal N-acetylglucosamine-6-O-sulfotransferase; MCDC1; mIGn6ST; N-acetylglucosamine 6-O-sulfotransferase 3 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
23563 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9GZS9 | |
CHST5 | |
CHO-derived recombinant human Carbohydrate Sulfotransferase5/CHST5 Ser28-Arg390 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |