missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA583721
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Immunogen sequence: PKRKMRRRSS CSSEPNTPKS AKCEGDIFTF DRTGTEAEDV LGELEYDKVP YSSLRRTLDQ RRALVMQLFQ DHGFFPSAQA TAAFQARYAD IFPSKVCLQL KIREVRQKIM QA
This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.Spezifikation
CIC | |
Polyclonal | |
Unconjugated | |
CIC | |
1200010B10Rik, Capicua, capicua homolog, capicua homolog (Drosophila), capicua transcriptional repressor, CIC, KIAA0306, mKIAA0306, Protein capicua homolog, protein capicua homolog, LOW QUALITY PROTEIN: protein capicua homolog | |
Rabbit | |
Anitgen affinity chromatography, Antigen affinity chromatography | |
RUO | |
23152 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide, pH 7.2 | |
Q96RK0 | |
CIC | |
Recombinant protein corresponding to Human CIC | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |