missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CID755673, MedChemExpress
Description
CID755673 is a potent PKD inhibitor with IC50s of 182 nM, 280 nM and 227 nM for PKD1, PKD2 and PKD3, respectively.
Specifications
Specifications
| Chemischer Name oder Material | CID755673 |
| CAS | 521937-07-5 |
| Farbe | Off-White |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C12H11NO3 |
| Menge | 10 mg |
| Löslichkeitsinformationen | DMSO : 100 mg/mL (460.36 mM; Need ultrasonic) |
| SMILES | O=C1NCCCC2=C1OC3=CC=C(O)C=C32 |
| Molekulargewicht (g/mol) | 217.22 |
| Formelmasse | 217.22 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?