Learn More
STEMCELL Technologies CloneR™, 5 Pack
Artikelnummer. 15795593
Encompass
Ansicht ändern
Click to view available options
Menge:
5 x 10 ml
Packungsgröße:
5 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15795593 5 x 10 ml 5 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15795593 Lieferant STEMCELL Technologies Lieferanten-Nr. 05889
Please to purchase this item. Need a web account? Register with us today!
Defined supplement for single-cell cloning of human ES and iPS cells
  • CloneR™ is a defined, serum-free supplement designed to increase the cloning efficiency and single-cell survival of human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPS cells)
  • CloneR™ enables the robust generation of clonal cell lines without single-cell adaptation, thus minimizing the risk of acquiring genetic abnormalities
  • CloneR™ is compatible with the TeSR™ family of media for human ES and iPS cell maintenance as well as your choice of cell culture matrix
Spezifikation
Zelltyp Pluripotent Stem Cells
Zur Verwendung mit (Anwendung) Zellkultur
Menge 5 x 10 ml