Learn More
Lovibond™ COD VLR Tube Test, 2-60 mg/L
Each pack contains 25 COD vials, which are made of high-quality optical glass with a 16 mm diameter. Vials are packed in an EPS foam container which complies with global shipping and safety standards.
Marke: Lovibond™ 2423100
Weitere Details : CAS : 7664-93-9 Netto-Gewicht : 0.10000kg
Beschreibung
- Chemical Oxygen Demand (COD) is a common water quality parameter for municipal wastewater and industrial wastewater applications. It is used to quantify the amount of organics in a water sample and indicates the amount of oxygen that is available to be consumed by reactions with that solution.
- As results of a COD test are easy to reproduce and are available in two hours, it is widely used as a process control parameter. COD values give a good indication of what is currently happening in a system and over time it can be used to make predictions to further optimize the treatment process.
- Barcoded vial for easy method selection when used with Lovibond Spectrophotometers
- Pre-filled tubes reduce sample preparation and handling
- Part of the complete range of COD products from Lovibond™
Spezifikation
Chemical Oxygen Demand (COD) | |
Industrial Wastewater, Environmental Compliance, Municipal Wastewater | |
24 and 1 reagent blank | |
2 to 60 mg/L | |
Dichromate / H2SO4 |
Tube Test / Vial Test | |
Lovibond Spectrophotometers and Colorimeters | |
25 Tubes Tests | |
Lovibond RD125 Reactor | |
Sample prep and 2 hour digestion required |