Learn More
Hanna Instruments™ Leitfähigkeitslösung
Catalog Number 10117240
Premium quality 84 µS/cm conductivity standard that is prepared against a NIST traceable potassium chloride solution.
Typ: Standard 84 μS/cm, 460mL Flasche