missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CSTA (Human) Recombinant Protein
Human CSTA full-length ORF ( AAH10379, 1 a.a. - 98 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00001475-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: MIPGGLSEAKPATPEIQEIVDKVKPQLEEKTNETYGKLEAVQYKTQVVAGTNYYIKVRAGDNKYMHLKVFKSLPGQNEDLVLTGYQVDKNKDDELTGF
Spezifikation
AAH10379 | |
cystatin A (stefin A) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CSTA | |
GST |
1475 | |
Wheat germ expression system | |
25 μg | |
STF1, STFA | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |