Learn More
Invitrogen™ Cu/ZnSOD Human Matched Antibody Pair
Matched Antibody Pair
Marke: Invitrogen™ BMS222MST
Weitere Details : Netto-Gewicht : 0.23750kg
Beinhaltet:
- 1 vial (2.75mL) monoclonal Coating Antibody to human Cu/ZnSOD (100μg/mL)
- 1 vial (55μL) HRP-Conjugate anti-human Cu/ZnSOD monoclonal antibody
- 1 vial (60μL) human Cu/ZnSOD Standard protein, 400ng/mL
Beschreibung
The Human Cu/ZnSOD Matched Antibody Pair Kit comes with pre-matched antibody pairs and standards to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human Cu/ZnSOD. Buffer reagents and microplates needed to complete the ELISA reaction are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate,which is an overnight process. Samples,standards,or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody,a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
SOD1 (superoxide dismutase) binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. This isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene.Spezifikation
Cu/ZnSOD | |
Monoclonal | |
P00441 | |
SOD1 | |
RUO | |
6647 | |
-20°C |
ELISA | |
HRP | |
Cu/Zn Superoxide Dismutase | |
10 x 96 Tests | |
Primary | |
Human | |
ELISA Kit |
For Research Use Only.