Learn More
Ibidi Culture-Insert 2 Well in μ-Dish 35 mm, High ibiTreat
Artikelnummer. 17246223
Encompass_Preferred
Ansicht ändern
Click to view available options
Anzahl Wells:
2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Anzahl Wells
17246223 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17246223 Lieferant Ibidi Lieferanten-Nr. 81176
Please to purchase this item. Need a web account? Register with us today!
Culture-Insert 2 Well in μ-Dish 35 mm, High ibiTreat is a 2 well silicone insert with a defined cell-free gap of 500 μm.
  • A 2 well silicone insert with a defined cell-free gap of 500 μm
  • suitable for wound healing
  • migration assays
  • 2D invasion assays
  • and co-cultivation of cells
  • in a μ-Dish 35 mm high with #1.5 Polymer Coverslip bottom
  • tissue culture treated
  • sterilized
Spezifikation
Typ Kultureinsatz
Menge 30
Wells 2
Sterilität Steril
Anzahl Wells 2