missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL14 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10468-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene belongs to the cytokine gene family which encode secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC (Cys-X-Cys) subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes but not for lymphocytes, dendritic cells, neutrophils or macrophages. It has been implicated that this cytokine is involved in the homeostasis of monocyte-derived macrophages rather than in inflammation.Spezifikation
CXCL14 | |
Polyclonal | |
Unconjugated | |
CXCL14 | |
BMAC, bolekine, BRAK, C X C motif chemokine 14, Chemokine BRAK, CXCL14, Kec, KS1, MIP 2g, MIP2G, NJAC, PSEC0212, SCYB14, Small inducible cytokine B14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9547 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95715 | |
CXCL14 | |
CXCL14 Fusion Protein Ag0747 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |